Search Thermo Fisher Scientific
GACAAATTACCTTTTTAAGCCTAAA[A/G]AAATATAAATTCCTAACATGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607813 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPPR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLPPR4 - phospholipid phosphatase related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166252.1 | Intron | NP_001159724.1 | ||||
NM_014839.4 | Intron | NP_055654.2 | ||||
XM_011542498.2 | Intron | XP_011540800.1 |
Set Membership: |
HapMap |