Search
Search

ALDH1A3
LOC101927751
LRRK1GGCCGCCGAGGGGACGCCTCGCGAC[G/T]GTTCCTGGGAGAGCTGGCGGCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH1A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ALDH1A3 - aldehyde dehydrogenase 1 family member A3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC101927751 - uncharacterized LOC101927751 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LRRK1 - leucine rich repeat kinase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024652.4 | 226 | UTR 5 | NP_078928.3 | |||
| XM_005254979.3 | 226 | Intron | XP_005255036.1 | |||
| XM_011522012.2 | 226 | Intron | XP_011520314.1 | |||
| XM_011522013.2 | 226 | Intron | XP_011520315.1 | |||
| XM_011522014.2 | 226 | UTR 5 | XP_011520316.1 | |||
| XM_011522015.2 | 226 | Intron | XP_011520317.1 | |||
| XM_011522016.2 | 226 | Intron | XP_011520318.1 | |||
| XM_011522017.2 | 226 | Intron | XP_011520319.1 | |||
| XM_011522018.2 | 226 | Intron | XP_011520320.1 | |||
| XM_011522019.2 | 226 | UTR 5 | XP_011520321.1 | |||
| XM_017022570.1 | 226 | Intron | XP_016878059.1 | |||