Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600380 MIM: 174761 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR1H2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NR1H2 - nuclear receptor subfamily 1 group H member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLD1 - DAN polymerase delta 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256849.1 | Intron | NP_001243778.1 | ||||
NM_001308632.1 | Intron | NP_001295561.1 | ||||
NM_002691.3 | Intron | NP_002682.2 | ||||
XM_005259008.4 | Intron | XP_005259065.1 | ||||
XM_011527038.1 | Intron | XP_011525340.1 | ||||
XM_017026881.1 | Intron | XP_016882370.1 | ||||
XM_017026882.1 | Intron | XP_016882371.1 |
Set Membership: |
HapMap |