Search
Search

CYP2D6
LOC102723722
NDUFA6-AS1GGGTCCCAGGTCATCCTGTGCTC[-/AGTT]AGCAGCTCATCCAGCTGGGTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||
Literature Links: |
CYP2D6 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|
||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |
| CYP2D6 - cytochrome P450 family 2 subfamily D member 6 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000106.5 | 702 | Silent Mutation | NP_000097.3 | |||
| NM_001025161.2 | 702 | Silent Mutation | NP_001020332.2 | |||
| LOC102723722 - uncharacterized LOC102723722 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NDUFA6-AS1 - NDUFA6 antisense RNA 1 (head to head) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
Set Membership: |
|