Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGGGCTGGTCTTCTAAGGGCATCTT[A/G]GATCCGGGCCCTGGTGATTCCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605008 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADAMTS6 - ADAM metallopeptidase with thrombospondin type 1 motif 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_197941.3 | 4547 | UTR 3 | NP_922932.2 | |||
XM_011543113.2 | 4547 | UTR 3 | XP_011541415.1 | |||
XM_011543114.2 | 4547 | UTR 3 | XP_011541416.1 | |||
XM_011543115.2 | 4547 | UTR 3 | XP_011541417.1 | |||
XM_011543116.2 | 4547 | UTR 3 | XP_011541418.1 | |||
XM_011543117.1 | 4547 | UTR 3 | XP_011541419.1 | |||
XM_011543119.2 | 4547 | Intron | XP_011541421.1 | |||
XM_011543120.2 | 4547 | Intron | XP_011541422.1 | |||
XM_011543121.2 | 4547 | UTR 3 | XP_011541423.2 | |||
XM_011543122.2 | 4547 | Intron | XP_011541424.1 | |||
XM_011543123.2 | 4547 | UTR 3 | XP_011541425.1 | |||
XM_011543124.2 | 4547 | UTR 3 | XP_011541426.1 | |||
XM_011543125.1 | 4547 | UTR 3 | XP_011541427.1 | |||
XM_011543126.2 | 4547 | Intron | XP_011541428.1 | |||
XM_011543127.2 | 4547 | Intron | XP_011541429.1 |
Set Membership: |
HapMap |