Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
TAACATTTGAAAAACTATTTGGTCT[A/G]CCTGCACTGCAAACTTATAGATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608307 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPS1 - carbamoyl-phosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122633.2 | Intron | NP_001116105.1 | ||||
NM_001122634.3 | Intron | NP_001116106.1 | ||||
NM_001875.4 | Intron | NP_001866.2 | ||||
XM_011510640.1 | Intron | XP_011508942.1 | ||||
XM_011510641.2 | Intron | XP_011508943.1 | ||||
XM_011510642.2 | Intron | XP_011508944.1 | ||||
XM_011510643.2 | Intron | XP_011508945.1 | ||||
XM_011510644.2 | Intron | XP_011508946.1 |
Set Membership: |
HapMap |