Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTAATTGTAATGCCTGCTACA[A/G]AGCACTCTGTGAAAATACAAACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601275 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPM6A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPM6A - glycoprotein M6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261447.1 | 3076 | UTR 3 | NP_001248376.1 | |||
NM_001261448.1 | 3076 | UTR 3 | NP_001248377.1 | |||
NM_005277.4 | 3076 | UTR 3 | NP_005268.1 | |||
NM_201591.2 | 3076 | UTR 3 | NP_963885.1 | |||
NM_201592.2 | 3076 | UTR 3 | NP_963886.1 | |||
XM_006714183.1 | 3076 | UTR 3 | XP_006714246.1 | |||
XM_011531877.1 | 3076 | UTR 3 | XP_011530179.1 | |||
XM_017008036.1 | 3076 | UTR 3 | XP_016863525.1 |
Set Membership: |
HapMap |