Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGAAAAGTGTAGTTTTGAAGAAGCA[C/T]GAGAAGTTTTTGAAAACACTGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300746 | ||||||||||||||||||||
Literature Links: |
F9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
F9 - coagulation factor IX | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000133.3 | 268 | Nonsense Mutation | CGA,TGA | R,* 75 | NP_000124.1 | |
NM_001313913.1 | 268 | Nonsense Mutation | CGA,TGA | R,* 75 | NP_001300842.1 | |
XM_005262397.4 | 268 | Nonsense Mutation | CGA,TGA | R,* 75 | XP_005262454.1 |