Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCAGGAACTGTATGAGAAAGGA[A/C]AGCAGTCAGTTTTTCCTTTGAGGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609348 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REEP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
REEP3 - receptor accessory protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001330.2 | Intron | NP_001001330.1 | ||||
XM_011539501.2 | Intron | XP_011537803.1 | ||||
XM_017015896.1 | Intron | XP_016871385.1 |
Set Membership: |
HapMap |