Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGATAAAATACACTGCTGTCTG[G/T]TAAGAGCTTACAGGAAGGAAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603503 MIM: 609277 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADNP-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADNP-AS1 - ADNP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DPM1 - dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317034.1 | Intron | NP_001303963.1 | ||||
NM_001317035.1 | Intron | NP_001303964.1 | ||||
NM_001317036.1 | Intron | NP_001303965.1 | ||||
NM_003859.2 | Intron | NP_003850.1 | ||||
XM_011529093.2 | Intron | XP_011527395.1 |
MOCS3 - molybdenum cofactor synthesis 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |