Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GAAATGAGACTGGCATGCTTATCAC[C/T]AAACTAGAGACTTGTGATAAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AOAH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AOAH - acyloxyacyl hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177506.1 | Intron | NP_001170977.1 | ||||
NM_001177507.1 | Intron | NP_001170978.1 | ||||
NM_001637.3 | Intron | NP_001628.1 | ||||
XM_011515333.2 | Intron | XP_011513635.1 | ||||
XM_011515334.2 | Intron | XP_011513636.1 | ||||
XM_011515335.2 | Intron | XP_011513637.1 | ||||
XM_011515336.2 | Intron | XP_011513638.1 | ||||
XM_011515338.2 | Intron | XP_011513640.1 | ||||
XM_011515339.2 | Intron | XP_011513641.1 | ||||
XM_011515340.2 | Intron | XP_011513642.1 | ||||
XM_011515341.2 | Intron | XP_011513643.1 | ||||
XM_011515342.2 | Intron | XP_011513644.1 | ||||
XM_017012102.1 | Intron | XP_016867591.1 | ||||
XM_017012103.1 | Intron | XP_016867592.1 | ||||
XM_017012104.1 | Intron | XP_016867593.1 | ||||
XM_017012105.1 | Intron | XP_016867594.1 | ||||
XM_017012106.1 | Intron | XP_016867595.1 |
Set Membership: |
HapMap |