Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TATAAATCTCATGAGCACCCACAGC[C/G]GTCTACTACCATCGCTGGAATTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615117 MIM: 601788 | ||||||||||||||||||||
Literature Links: |
C2orf88 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf88 - chromosome 2 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042519.1 | 1244 | Intron | NP_001035984.1 | |||
NM_001042520.1 | 1244 | Intron | NP_001035985.1 | |||
NM_001042521.1 | 1244 | Intron | NP_001035986.1 | |||
NM_032321.2 | 1244 | Intron | NP_115697.2 | |||
XM_005246905.1 | 1244 | Intron | XP_005246962.1 | |||
XM_006712795.2 | 1244 | Intron | XP_006712858.1 | |||
XM_011511982.1 | 1244 | Intron | XP_011510284.1 | |||
XM_011511983.1 | 1244 | Intron | XP_011510285.1 | |||
XM_011511984.2 | 1244 | Intron | XP_011510286.1 | |||
XM_011511985.2 | 1244 | Intron | XP_011510287.1 | |||
XM_011511986.2 | 1244 | Intron | XP_011510288.1 | |||
XM_017005096.1 | 1244 | Intron | XP_016860585.1 |
MSTN - myostatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005259.2 | 1244 | Missense Mutation | CGC,GGC | R,G 371 | NP_005250.1 |