Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
CCAAGACGAGGAGTGAGAACAGCCT[A/C]GTACCTTTTACAGTGTGCACTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSRA - methionine sulfoxide reductase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135670.2 | Intron | NP_001129142.1 | ||||
NM_001135671.2 | Intron | NP_001129143.1 | ||||
NM_001199729.2 | Intron | NP_001186658.1 | ||||
NM_012331.4 | Intron | NP_036463.1 | ||||
XM_011543822.1 | Intron | XP_011542124.1 | ||||
XM_011543823.2 | Intron | XP_011542125.1 | ||||
XM_017013448.1 | Intron | XP_016868937.1 | ||||
XM_017013449.1 | Intron | XP_016868938.1 | ||||
XM_017013450.1 | Intron | XP_016868939.1 | ||||
XM_017013451.1 | Intron | XP_016868940.1 | ||||
XM_017013452.1 | Intron | XP_016868941.1 | ||||
XM_017013453.1 | Intron | XP_016868942.1 | ||||
XM_017013454.1 | Intron | XP_016868943.1 |