Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ACAGTGATGTGGATCAGCTTGCTGG[A/C]CAGGTGCTCCAGGCTGTCCTTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615956 MIM: 616743 | ||||||||||||||||||||
Literature Links: |
CCDC151 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC151 - coiled-coil domain containing 151 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302453.1 | 1551 | Missense Mutation | GCC,TCC | A,S 416 | NP_001289382.1 | |
NM_001302454.1 | 1551 | Missense Mutation | GCC,TCC | A,S 410 | NP_001289383.1 | |
NM_145045.4 | 1551 | Missense Mutation | GCC,TCC | A,S 470 | NP_659482.3 | |
XM_017026241.1 | 1551 | Intron | XP_016881730.1 |
RGL3 - ral guanine nucleotide dissociation stimulator like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |