Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAATCCTAATCCTCTTTGAGGGCTA[A/G]TCTAAGATTCTTCTCATTCCTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604587 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCOCO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CALCOCO2 - calcium binding and coiled-coil domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261390.1 | Intron | NP_001248319.1 | ||||
NM_001261391.1 | Intron | NP_001248320.1 | ||||
NM_001261393.1 | Intron | NP_001248322.1 | ||||
NM_001261395.1 | Intron | NP_001248324.1 | ||||
NM_005831.4 | Intron | NP_005822.1 |