Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTGCCATCCTCTTTCACGTCT[C/T]TAAGTCTTTGCATATACTGTATCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616711 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAOK3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TAOK3 - TAO kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016281.3 | Intron | NP_057365.3 | ||||
XM_005253897.1 | Intron | XP_005253954.1 | ||||
XM_005253898.1 | Intron | XP_005253955.1 | ||||
XM_006719445.2 | Intron | XP_006719508.1 | ||||
XM_011538437.1 | Intron | XP_011536739.1 | ||||
XM_011538439.1 | Intron | XP_011536741.1 | ||||
XM_017019408.1 | Intron | XP_016874897.1 | ||||
XM_017019409.1 | Intron | XP_016874898.1 | ||||
XM_017019410.1 | Intron | XP_016874899.1 | ||||
XM_017019411.1 | Intron | XP_016874900.1 | ||||
XM_017019412.1 | Intron | XP_016874901.1 | ||||
XM_017019413.1 | Intron | XP_016874902.1 |
Set Membership: |
HapMap |