Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCOC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| SCOC - short coiled-coil protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001153446.1 | Intron | NP_001146918.1 | ||||
| NM_001153484.1 | Intron | NP_001146956.1 | ||||
| NM_001153552.1 | Intron | NP_001147024.1 | ||||
| NM_001153585.1 | Intron | NP_001147057.1 | ||||
| NM_001153635.1 | Intron | NP_001147107.1 | ||||
| NM_001153663.1 | Intron | NP_001147135.1 | ||||
| NM_001153690.1 | Intron | NP_001147162.1 | ||||
| NM_032547.2 | Intron | NP_115936.2 | ||||