Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||
Literature Links: |
GSTM1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |
| GSTM1 - glutathione S-transferase mu 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000561.3 | Intron | NP_000552.2 | ||||
| NM_146421.2 | Intron | NP_666533.1 | ||||
| XM_005270782.4 | Intron | XP_005270839.1 | ||||
| GSTM2 - glutathione S-transferase mu 2 (muscle) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
Set Membership: |
|