Search Thermo Fisher Scientific
GTGGACAGATTGGAATAATATGTCC[A/C]AGTTTTTCCAGTATGGATGGCGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf1 - chromosome 11 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022761.2 | 318 | Missense Mutation | AAG,CAG | K,Q 49 | NP_073598.1 | |
XM_011542957.2 | 318 | Missense Mutation | AAG,CAG | K,Q 65 | XP_011541259.1 | |
XM_011542958.2 | 318 | Missense Mutation | AAG,CAG | K,Q 65 | XP_011541260.1 | |
XM_017018129.1 | 318 | Missense Mutation | AAG,CAG | K,Q 49 | XP_016873618.1 |
FDXACB1 - ferredoxin-fold anticodon binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |