Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
PERM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PERM1 - PPARGC1 and ESRR induced regulator, muscle 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001291366.1 | 2048 | Missense Mutation | GCG,GTG | A,V 777 | NP_001278295.1 | |
| NM_001291367.1 | 2048 | Missense Mutation | GCG,GTG | A,V 683 | NP_001278296.1 | |
| XM_017002583.1 | 2048 | Missense Mutation | GCG,GTG | A,V 777 | XP_016858072.1 | |
| XM_017002584.1 | 2048 | Missense Mutation | GCG,GTG | A,V 777 | XP_016858073.1 | |
| XM_017002585.1 | 2048 | Missense Mutation | GCG,GTG | A,V 755 | XP_016858074.1 | |
| PLEKHN1 - pleckstrin homology domain containing N1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||