Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGCTCAGAGCAAGCCAATAAATACA[C/G]GGGGAGTGAGTTGAGGCTTTAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609772 | ||||||||||||||||||||
Literature Links: |
CTTNBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTTNBP2 - cortactin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033427.2 | Intron | NP_219499.1 | ||||
XM_005250634.4 | Intron | XP_005250691.1 | ||||
XM_005250635.3 | Intron | XP_005250692.1 | ||||
XM_011516615.2 | Intron | XP_011514917.1 | ||||
XM_011516616.2 | Intron | XP_011514918.1 | ||||
XM_011516618.2 | Intron | XP_011514920.1 | ||||
XM_017012706.1 | Intron | XP_016868195.1 | ||||
XM_017012707.1 | Intron | XP_016868196.1 | ||||
XM_017012708.1 | Intron | XP_016868197.1 |