Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ATATTTTTCCATTCATATCTTTGTA[C/T]TTTTAAATAAAAGTTAAATCATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAMD12 - sterile alpha motif domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101676.1 | Intron | NP_001095146.1 | ||||
NM_207506.2 | Intron | NP_997389.2 | ||||
XM_011517031.2 | Intron | XP_011515333.1 | ||||
XM_011517034.2 | Intron | XP_011515336.1 | ||||
XM_011517035.2 | Intron | XP_011515337.1 | ||||
XM_017013409.1 | Intron | XP_016868898.1 | ||||
XM_017013410.1 | Intron | XP_016868899.1 | ||||
XM_017013411.1 | Intron | XP_016868900.1 | ||||
XM_017013412.1 | Intron | XP_016868901.1 |