Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGCCTCTGTTTACCTCGGTCGG[A/G]GCGGCCCCGGGGTGGGGCGGGGCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM65A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM65A - family with sequence similarity 65 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193522.1 | Intron | NP_001180451.1 | ||||
NM_001193523.1 | Intron | NP_001180452.1 | ||||
NM_001193524.1 | Intron | NP_001180453.1 | ||||
NM_024519.3 | Intron | NP_078795.2 | ||||
XM_011523321.1 | Intron | XP_011521623.1 | ||||
XM_011523322.1 | Intron | XP_011521624.1 | ||||
XM_011523324.2 | Intron | XP_011521626.1 | ||||
XM_011523325.1 | Intron | XP_011521627.1 | ||||
XM_011523326.1 | Intron | XP_011521628.1 | ||||
XM_017023663.1 | Intron | XP_016879152.1 | ||||
XM_017023664.1 | Intron | XP_016879153.1 |
LOC100505942 - uncharacterized LOC100505942 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |