Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGGCCACCATCGCCAAGAACC[A/G]GAACCTGCACTCACCCATGTACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155555 MIM: 612326 MIM: 602661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MC1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MC1R - melanocortin 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002386.3 | 1580 | Missense Mutation | CAG,CGG | Q,R 67 | NP_002377.4 |
TCF25 - transcription factor 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBB3 - tubulin beta 3 class III | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |