Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGCGAGGCAGCTGGGGCAGGA[A/G]CAAGGCTGGAAATGCGTTCACTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109610 | ||||||||||||||||||||
Literature Links: |
TSPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPO - translocator protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000714.5 | Intron | NP_000705.2 | ||||
NM_001256530.1 | Intron | NP_001243459.1 | ||||
NM_001256531.1 | Intron | NP_001243460.1 |
TTLL12 - tubulin tyrosine ligase like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |