Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGCAGTTCCTCACTATCCTGCCCCG[C/T]GAGTTCTACGCCTGGATCCGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GS1-259H13.2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GS1-259H13.2 - transmembrane protein 225-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | NP_660090.2 | |
XM_005250194.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_005250251.1 | |
XM_011515905.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_011514207.1 | |
XM_011515906.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_011514208.1 | |
XM_011515907.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_011514209.1 | |
XM_011515908.2 | 870 | UTR 5 | XP_011514210.1 | |||
XM_011515909.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_011514211.1 | |
XM_011515910.2 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_011514212.1 | |
XM_017011824.1 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_016867313.1 | |
XM_017011825.1 | 870 | UTR 5 | XP_016867314.1 | |||
XM_017011826.1 | 870 | UTR 5 | XP_016867315.1 | |||
XM_017011827.1 | 870 | Silent Mutation | CGC,CGT | R,R 96 | XP_016867316.1 |