Search
Search

KRT40
KRTAP3-2
KRTAP3-3AGGTGCTGGGCAGGCAGACTCCACA[A/G]CGGCAGGATTTGTCAGAGGAGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
KRT40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| KRT40 - keratin 40 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| KRTAP3-2 - keratin associated protein 3-2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| KRTAP3-3 - keratin associated protein 3-3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_033185.2 | 117 | Silent Mutation | CGC,CGT | R,R 27 | NP_149441.1 | |