Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTTGTATCATAAATGAAACTT[C/T]AGACTGGTTAAAGAAAATTGAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APP - amyloid beta precursor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000484.3 | 3369 | UTR 3 | NP_000475.1 | |||
NM_001136016.3 | 3369 | UTR 3 | NP_001129488.1 | |||
NM_001136129.2 | 3369 | UTR 3 | NP_001129601.1 | |||
NM_001136130.2 | 3369 | UTR 3 | NP_001129602.1 | |||
NM_001136131.2 | 3369 | UTR 3 | NP_001129603.1 | |||
NM_001204301.1 | 3369 | UTR 3 | NP_001191230.1 | |||
NM_001204302.1 | 3369 | UTR 3 | NP_001191231.1 | |||
NM_001204303.1 | 3369 | UTR 3 | NP_001191232.1 | |||
NM_201413.2 | 3369 | UTR 3 | NP_958816.1 | |||
NM_201414.2 | 3369 | UTR 3 | NP_958817.1 |