Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGAGAAAGGAATGGACAAAATAAAT[A/G]ATTAAACAGGAGAGAATAAAATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPAL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIPAL3 - NIPA like domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322854.1 | Intron | NP_001309783.1 | ||||
NM_001322855.1 | Intron | NP_001309784.1 | ||||
NM_001322856.1 | Intron | NP_001309785.1 | ||||
NM_001322857.1 | Intron | NP_001309786.1 | ||||
NM_001322858.1 | Intron | NP_001309787.1 | ||||
NM_001322859.1 | Intron | NP_001309788.1 | ||||
NM_001322860.1 | Intron | NP_001309789.1 | ||||
NM_001322861.1 | Intron | NP_001309790.1 | ||||
NM_001322862.1 | Intron | NP_001309791.1 | ||||
NM_001322863.1 | Intron | NP_001309792.1 | ||||
NM_001322864.1 | Intron | NP_001309793.1 | ||||
NM_001322865.1 | Intron | NP_001309794.1 | ||||
NM_001322866.1 | Intron | NP_001309795.1 | ||||
NM_001322868.1 | Intron | NP_001309797.1 | ||||
NM_020448.4 | Intron | NP_065181.1 | ||||
XM_011541808.2 | Intron | XP_011540110.1 | ||||
XM_017001868.1 | Intron | XP_016857357.1 | ||||
XM_017001869.1 | Intron | XP_016857358.1 | ||||
XM_017001870.1 | Intron | XP_016857359.1 |
STPG1 - sperm tail PG-rich repeat containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |