Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GGGTGTTTAAAGACCAAAAGAAAAA[A/G]AACAGTCAGAGGCAGAATTCCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STX2 - syntaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001980.3 | 2890 | UTR 3 | NP_001971.2 | |||
NM_194356.2 | 2890 | UTR 3 | NP_919337.1 | |||
XM_005253557.1 | 2890 | UTR 3 | XP_005253614.1 | |||
XM_017018978.1 | 2890 | UTR 3 | XP_016874467.1 | |||
XM_017018979.1 | 2890 | UTR 3 | XP_016874468.1 | |||
XM_017018980.1 | 2890 | UTR 3 | XP_016874469.1 | |||
XM_017018981.1 | 2890 | Intron | XP_016874470.1 | |||
XM_017018982.1 | 2890 | UTR 3 | XP_016874471.1 | |||
XM_017018983.1 | 2890 | UTR 3 | XP_016874472.1 | |||
XM_017018984.1 | 2890 | UTR 3 | XP_016874473.1 | |||
XM_017018985.1 | 2890 | UTR 3 | XP_016874474.1 | |||
XM_017018986.1 | 2890 | Intron | XP_016874475.1 | |||
XM_017018987.1 | 2890 | Intron | XP_016874476.1 | |||
XM_017018988.1 | 2890 | Intron | XP_016874477.1 | |||
XM_017018989.1 | 2890 | Intron | XP_016874478.1 |