Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTGTGGCTAATCCCTTGGTCA[A/G]TGACCTCATTCATGGCAAAAATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF23 - kinesin family member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281301.1 | 462 | Missense Mutation | ATG,GTG | M,V 34 | NP_001268230.1 | |
NM_004856.6 | 462 | Missense Mutation | AAT,AGT | N,S 98 | NP_004847.2 | |
NM_138555.3 | 462 | Missense Mutation | AAT,AGT | N,S 98 | NP_612565.1 | |
XM_005254796.2 | 462 | Missense Mutation | AAT,AGT | N,S 98 | XP_005254853.1 | |
XM_005254797.2 | 462 | Missense Mutation | AAT,AGT | N,S 98 | XP_005254854.1 | |
XM_005254798.2 | 462 | Missense Mutation | AAT,AGT | N,S 98 | XP_005254855.1 | |
XM_005254799.2 | 462 | Missense Mutation | AAT,AGT | N,S 98 | XP_005254856.1 | |
XM_011522238.1 | 462 | Missense Mutation | ATG,GTG | M,V 34 | XP_011520540.1 | |
XM_011522239.2 | 462 | UTR 5 | XP_011520541.1 | |||
XM_011522240.1 | 462 | Intron | XP_011520542.1 | |||
XM_017022733.1 | 462 | Missense Mutation | AAT,AGT | N,S 98 | XP_016878222.1 |
LOC145694 - uncharacterized LOC145694 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |