Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGTGGTTTCCACTGCTGAGCCTAGT[A/G]AGGGGTGACTGAGAAGTTTCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRT4 - proline rich transmembrane protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114726.2 | Intron | NP_001108198.2 | ||||
NM_001174164.1 | Intron | NP_001167635.1 | ||||
XM_005250342.3 | Intron | XP_005250399.1 | ||||
XM_005250343.3 | Intron | XP_005250400.1 | ||||
XM_006715985.3 | Intron | XP_006716048.1 | ||||
XM_017012205.1 | Intron | XP_016867694.1 | ||||
XM_017012206.1 | Intron | XP_016867695.1 |