Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GAAAATTTCAGACAGTGTGACATTA[A/T]TTATTTGTCTTTGCACAGAATGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100670 MIM: 610413 | ||||||||||||||||||||
Literature Links: |
ALDH1B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH1B1 - aldehyde dehydrogenase 1 family member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGFBPL1 - insulin like growth factor binding protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007563.2 | 3296 | UTR 3 | NP_001007564.1 | |||
XM_017014699.1 | 3296 | Intron | XP_016870188.1 |