Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCTGGGACTCTGTGCAAAGCC[C/T]CCTGGGAATTCTGGAACAACTAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605687 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL19 - interleukin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013371.3 | Intron | NP_037503.2 | ||||
NM_153758.2 | Intron | NP_715639.1 | ||||
XM_011509450.2 | Intron | XP_011507752.1 |