Search Thermo Fisher Scientific
GTGGATCTCAGACGGCACACAGCTC[A/G]TAAGTCCTCCATCAAAGCTGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112264 MIM: 178620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP1 - bone morphogenetic protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199.3 | 505 | Missense Mutation | CAT,CGT | H,R 80 | NP_001190.1 | |
NM_006129.4 | 505 | Missense Mutation | CAT,CGT | H,R 80 | NP_006120.1 | |
XM_006716386.2 | 505 | Missense Mutation | CAT,CGT | H,R 80 | XP_006716449.2 | |
XM_011544617.1 | 505 | Missense Mutation | CAT,CGT | H,R 80 | XP_011542919.1 | |
XM_017013738.1 | 505 | Missense Mutation | CAT,CGT | H,R 80 | XP_016869227.1 |
SFTPC - surfactant protein C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |