Search
Search

INSIG1TCACGATTCTGAGGGAAAAAAATTC[C/T]GTATGGAGGTTTCTCTTTTGCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| INSIG1 - insulin induced gene 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_005542.4 | Intron | NP_005533.2 | ||||
| NM_198336.2 | Intron | NP_938150.2 | ||||
| NM_198337.2 | Intron | NP_938151.1 | ||||
| XM_005249542.1 | Intron | XP_005249599.1 | ||||
| XM_005249543.1 | Intron | XP_005249600.1 | ||||
| XM_017012177.1 | Intron | XP_016867666.1 | ||||