Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___1024375_10
          See other HLA-DMB GT Assays ›
          SNP ID:
          rs68600
          Gene
          HLA-DMB
          Gene Name
          major histocompatibility complex, class II, DM beta
          Set Membership:
          > HapMap > Validated
          Chromosome Location:
          Chr.6: 32935947 - 32935947 on Build GRCh38
          Polymorphism:
          C/T, Transition Substitution
          Context Sequence [VIC/FAM]:

          AGGCATCCTCTTTCCTTCCTTTACC[C/T]CAAAGCCACCCTTATCAGGATAAAG

          Assay ID C___1024375_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 142856

          Literature Links:

          HLA-DMB PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.39)
          (0.61)
          Caucasian
          C (0.45)
          (0.55)
          CEPH (CEU)
          A (0.48)
          (0.52)
          EAS
          A (0.40)
          (0.60)
          African American
          T (0.24)
          (0.76)
          YRI (Yoruba)
          A (0.33)
          (0.67)
          SAS
          A (0.41)
          (0.59)
          Chinese - Not Available JPT (Japanese)
          A (0.47)
          (0.53)
          AFR
          A (0.28)
          (0.72)
          Japanese - Not Available CHB (Han Chinese)
          A (0.42)
          (0.58)
          EUR
          A (0.47)
          (0.53)
          AMR
          A (0.44)
          (0.56)
          HLA-DMB - major histocompatibility complex, class II, DM beta
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_002118.4 Intron NP_002109.2

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs58879393] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          HapMap Validated

          Panther Classification:

          Molecular Function -

          defense/immunity protein major histocompatibility complex antigen

          Gene Ontology Categories:

          Function(s) Process(es)

          MHC class II protein complex assembly
          peptide antigen assembly with MHC class II protein complex
          immune response
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          positive regulation of T cell proliferation
          positive regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          MHC class II protein complex binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0