Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HFE PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| HFE - hemochromatosis | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000410.3 | 465 | Missense Mutation | TAC,TGC | Y,C 282 | NP_000401.1 | |
| NM_001300749.1 | 465 | Missense Mutation | TAC,TGC | Y,C 282 | NP_001287678.1 | |
| NM_139003.2 | 465 | Missense Mutation | TAC,TGC | Y,C 176 | NP_620572.1 | |
| NM_139004.2 | 465 | Missense Mutation | TAC,TGC | Y,C 190 | NP_620573.1 | |
| NM_139006.2 | 465 | Missense Mutation | TAC,TGC | Y,C 268 | NP_620575.1 | |
| NM_139007.2 | 465 | Missense Mutation | TAC,TGC | Y,C 194 | NP_620576.1 | |
| NM_139008.2 | 465 | Missense Mutation | TAC,TGC | Y,C 180 | NP_620577.1 | |
| NM_139009.2 | 465 | Missense Mutation | TAC,TGC | Y,C 259 | NP_620578.1 | |
| NM_139010.2 | 465 | Missense Mutation | TAC,TGC | Y,C 102 | NP_620579.1 | |
| NM_139011.2 | 465 | Intron | NP_620580.1 | |||
| XM_011514543.2 | 465 | Missense Mutation | TAC,TGC | Y,C 282 | XP_011512845.1 | |
Set Membership: |
|