Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| SLC2A9 - solute carrier family 2 member 9 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001001290.1 | Intron | NP_001001290.1 | ||||
| NM_020041.2 | Intron | NP_064425.2 | ||||
| XM_006713968.3 | Intron | XP_006714031.1 | ||||
| XM_006713969.2 | Intron | XP_006714032.1 | ||||
| XM_011513856.2 | Intron | XP_011512158.1 | ||||
| XM_011513857.1 | Intron | XP_011512159.1 | ||||
| XM_011513858.1 | Intron | XP_011512160.1 | ||||
| XM_011513859.2 | Intron | XP_011512161.1 | ||||
| XM_011513860.2 | Intron | XP_011512162.1 | ||||
| XM_011513861.2 | Intron | XP_011512163.1 | ||||
| XM_011513862.2 | Intron | XP_011512164.1 | ||||
| XM_011513863.2 | Intron | XP_011512165.1 | ||||
| XM_011513864.2 | Intron | XP_011512166.1 | ||||
| XM_011513865.2 | Intron | XP_011512167.1 | ||||
| XM_011513866.2 | Intron | XP_011512168.1 | ||||
| XM_011513867.2 | Intron | XP_011512169.1 | ||||
| XM_011513868.2 | Intron | XP_011512170.1 | ||||
| XM_017008457.1 | Intron | XP_016863946.1 | ||||
| XM_017008458.1 | Intron | XP_016863947.1 | ||||
| XM_017008459.1 | Intron | XP_016863948.1 | ||||
| XM_017008460.1 | Intron | XP_016863949.1 | ||||