Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600988 MIM: 611219 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDDC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HDDC3 - HD domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286451.1 | 838 | UTR 3 | NP_001273380.1 | |||
NM_198527.3 | 838 | UTR 3 | NP_940929.1 |
MAN2A2 - mannosidase alpha class 2A member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC45A - unc-45 myosin chaperone A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039675.1 | 838 | Intron | NP_001034764.1 | |||
NM_001323619.1 | 838 | Intron | NP_001310548.1 | |||
NM_001323620.1 | 838 | Intron | NP_001310549.1 | |||
NM_001323621.1 | 838 | Intron | NP_001310550.1 | |||
NM_018671.4 | 838 | Intron | NP_061141.2 | |||
XM_005254963.3 | 838 | Intron | XP_005255020.1 | |||
XM_011521779.2 | 838 | Intron | XP_011520081.1 | |||
XM_011521780.2 | 838 | Intron | XP_011520082.1 | |||
XM_017022407.1 | 838 | Intron | XP_016877896.1 |
Set Membership: |
HapMap Validated |