Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___1342401_10
          See other SLC28A1 GT Assays ›
          SNP ID:
          rs12901610
          Gene
          SLC28A1
          Gene Name
          solute carrier family 28 member 1
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.15: 84922642 - 84922642 on Build GRCh38
          Polymorphism:
          A/G, Transition substitution
          Context Sequence [VIC/FAM]:

          GAGTGACCCTTCAAACACACAAAGC[A/G]GATCATTCCACTCCTGCCTAAGCAA

          Assay ID C___1342401_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 606207

          Literature Links:

          SLC28A1 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.20)
          (0.80)
          Caucasian - Not Available CEPH (CEU)
          A (0.06)
          (0.94)
          EAS
          A (0.49)
          (0.51)
          African American - Not Available YRI (Yoruba)
          A (0.14)
          (0.86)
          SAS
          A (0.23)
          (0.77)
          Chinese - Not Available JPT (Japanese)
          A (0.49)
          (0.51)
          AFR
          A (0.10)
          (0.90)
          Japanese - Not Available CHB (Han Chinese)
          A (0.43)
          (0.57)
          EUR
          A (0.05)
          (0.95)
          AMR
          A (0.13)
          (0.87)
          SLC28A1 - solute carrier family 28 member 1
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001287761.1 Intron NP_001274690.1
          NM_001287762.1 Intron NP_001274691.1
          NM_001321721.1 Intron NP_001308650.1
          NM_001321722.1 Intron NP_001308651.1
          NM_004213.4 Intron NP_004204.3
          NM_201651.2 Intron NP_964014.1
          XM_011522203.2 Intron XP_011520505.1
          XM_011522204.2 Intron XP_011520506.1
          XM_011522205.2 Intron XP_011520507.1
          XM_011522206.2 Intron XP_011520508.1
          XM_011522207.2 Intron XP_011520509.1
          XM_011522208.2 Intron XP_011520510.1
          XM_011522209.2 Intron XP_011520511.1
          XM_011522210.2 Intron XP_011520512.1
          XM_011522211.2 Intron XP_011520513.1
          XM_011522212.1 Intron XP_011520514.1
          XM_011522214.2 Intron XP_011520516.1
          XM_011522215.2 Intron XP_011520517.1
          XM_011522216.2 Intron XP_011520518.1
          XM_011522217.1 Intron XP_011520519.1
          XM_011522218.2 Intron XP_011520520.1
          XM_017022723.1 Intron XP_016878212.1

          Back To Top

          More Information


          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          pyrimidine nucleobase transport
          nucleoside transport
          pyrimidine-containing compound transmembrane transport
          nucleoside transmembrane transport
          purine nucleobase transmembrane transport
          nucleoside transmembrane transporter activity
          nucleoside:sodium symporter activity
          pyrimidine- and adenine-specific:sodium symporter activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline