Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCEMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
| Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
| EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
| SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
| AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
| EUR - Not Available | ||||||||
| AMR - Not Available |
| MCEMP1 - mast cell expressed membrane protein 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| RETN - resistin | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001193374.1 | Intron | NP_001180303.1 | ||||
| NM_020415.3 | Intron | NP_065148.1 | ||||