Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C___1397673_10
          See other PDGFC GT Assays ›
          SNP ID:
          rs1425486
          Gene
          PDGFC
          Gene Name
          platelet derived growth factor C
          Set Membership:
          > HapMap > Validated
          Chromosome Location:
          Chr.4: 156762533 - 156762533 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          AAAAATCCAGTCTGTCCTATCAGCA[C/T]GACACAAGTTTAATTCATAGCAACA

          Assay ID C___1397673_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 608452

          Literature Links:

          PDGFC PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          C (0.47)
          (0.53)
          Caucasian - Not Available CEPH (CEU)
          T (0.34)
          (0.66)
          EAS
          T (0.32)
          (0.68)
          African American - Not Available YRI (Yoruba)
          C (0.12)
          (0.88)
          SAS
          C (0.37)
          (0.63)
          Japanese
          T (0.32)
          (0.68)
          JPT (Japanese)
          T (0.34)
          (0.66)
          AFR
          C (0.16)
          (0.84)
          Chinese
          T (0.28)
          (0.72)
          CHB (Han Chinese)
          T (0.34)
          (0.66)
          EUR
          T (0.36)
          (0.64)
          AMR
          T (0.37)
          (0.63)
          PDGFC - platelet derived growth factor C
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_016205.2 1859 UTR 3 NP_057289.1
          XM_011532124.2 1859 UTR 3 XP_011530426.1
          XM_017008455.1 1859 UTR 3 XP_016863944.1
          XM_017008456.1 1859 UTR 3 XP_016863945.1

          Back To Top

          More Information


          Set Membership:

          HapMap Validated

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of transmembrane receptor protein tyrosine kinase activity
          central nervous system development
          blood coagulation
          embryo development
          organ morphogenesis
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of cell migration
          positive regulation of protein autophosphorylation
          positive regulation of MAP kinase activity
          positive regulation of DNA replication
          platelet-derived growth factor receptor signaling pathway
          positive regulation of fibroblast proliferation
          digestive tract development
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell division
          bone development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          platelet-derived growth factor receptor binding
          protein binding
          growth factor activity
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0