Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CCTCGCGTCGAGGTTTCCATCCCTG[G/T]CTCTCAGCATTGCCCGTCAGCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612034 MIM: 180535 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
APC2 - APC2, WNT signaling pathway regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005883.2 | Intron | NP_005874.1 | ||||
XM_005259475.2 | Intron | XP_005259532.1 | ||||
XM_006722607.2 | Intron | XP_006722670.1 | ||||
XM_006722608.3 | Intron | XP_006722671.2 | ||||
XM_006722609.3 | Intron | XP_006722672.1 | ||||
XM_006722610.3 | Intron | XP_006722673.2 |
RPS15 - ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |