Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
ATTAAGAAAGCACAAAGCATTTTGG[G/T]CATGCTAAGGAAGGAGAAAGTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNOT6 - CCR4-NOT transcription complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303241.1 | Intron | NP_001290170.1 | ||||
XM_005265953.1 | Intron | XP_005266010.1 | ||||
XM_011534605.2 | Intron | XP_011532907.1 | ||||
XM_011534606.1 | Intron | XP_011532908.1 | ||||
XM_011534607.1 | Intron | XP_011532909.1 | ||||
XM_017009670.1 | Intron | XP_016865159.1 | ||||
XM_017009671.1 | Intron | XP_016865160.1 | ||||
XM_017009672.1 | Intron | XP_016865161.1 |