Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGGATCACTTGAGGCCAAAGATTCG[A/G]GACCAGACCGGGCAACATAGTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRTC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DMRTC2 - DMRT like family C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYPD4 - LY6/PLAUR domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291419.1 | Intron | NP_001278348.1 | ||||
NM_173506.6 | Intron | NP_775777.3 | ||||
XM_005258543.3 | Intron | XP_005258600.1 | ||||
XM_005278383.4 | Intron | XP_005278440.1 | ||||
XM_005278385.4 | Intron | XP_005278442.1 | ||||
XM_005278387.4 | Intron | XP_005278444.1 | ||||
XM_005278388.3 | Intron | XP_005278445.1 | ||||
XM_006723035.2 | Intron | XP_006723098.1 | ||||
XM_011526483.2 | Intron | XP_011524785.1 | ||||
XM_011526484.2 | Intron | XP_011524786.1 |