Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAATGTGGTCATTAATTAAAGG[C/T]TGTAGGCAGATCTTTTTCTATCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604698 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AKAP12 - A-kinase anchoring protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005100.3 | Intron | NP_005091.2 | ||||
NM_144497.2 | Intron | NP_653080.1 | ||||
XM_005267235.2 | Intron | XP_005267292.1 | ||||
XM_017011517.1 | Intron | XP_016867006.1 | ||||
XM_017011518.1 | Intron | XP_016867007.1 |
Set Membership: |
HapMap Validated |