Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| KLHL24 - kelch like family member 24 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_017644.3 | Intron | NP_060114.2 | ||||
| XM_005247552.2 | Intron | XP_005247609.1 | ||||
| XM_005247553.1 | Intron | XP_005247610.1 | ||||
| XM_005247554.2 | Intron | XP_005247611.1 | ||||
| XM_005247555.1 | Intron | XP_005247612.1 | ||||
| XM_005247556.1 | Intron | XP_005247613.1 | ||||
| XM_011512938.2 | Intron | XP_011511240.1 | ||||
| XM_017006650.1 | Intron | XP_016862139.1 | ||||
| XM_017006651.1 | Intron | XP_016862140.1 | ||||
| XM_017006652.1 | Intron | XP_016862141.1 | ||||
| XM_017006653.1 | Intron | XP_016862142.1 | ||||
| XM_017006654.1 | Intron | XP_016862143.1 | ||||
| XM_017006655.1 | Intron | XP_016862144.1 | ||||
| XM_017006656.1 | Intron | XP_016862145.1 | ||||
| XM_017006657.1 | Intron | XP_016862146.1 | ||||
| XM_017006658.1 | Intron | XP_016862147.1 | ||||
| XM_017006659.1 | Intron | XP_016862148.1 | ||||
| XM_017006660.1 | Intron | XP_016862149.1 | ||||
| XM_017006661.1 | Intron | XP_016862150.1 | ||||
| XM_017006662.1 | Intron | XP_016862151.1 | ||||
| XM_017006663.1 | Intron | XP_016862152.1 | ||||