Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3TC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| SH3TC1 - SH3 domain and tetratricopeptide repeats 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001318480.1 | Intron | NP_001305409.1 | ||||
| NM_018986.4 | Intron | NP_061859.3 | ||||
| XM_006713889.2 | Intron | XP_006713952.1 | ||||
| XM_006713891.3 | Intron | XP_006713954.1 | ||||
| XM_011513488.1 | Intron | XP_011511790.1 | ||||
| XM_011513489.2 | Intron | XP_011511791.1 | ||||
| XM_017008300.1 | Intron | XP_016863789.1 | ||||
| XM_017008301.1 | Intron | XP_016863790.1 | ||||
| XM_017008302.1 | Intron | XP_016863791.1 | ||||