Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AAGCCTGAGATTTGGTTAGGAGATA[A/G]AAGAAAATCTTAAAAAGCGTGGTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
GRAMD1C PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GRAMD1C - GRAM domain containing 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172105.1 | Intron | NP_001165576.1 | ||||
NM_017577.4 | Intron | NP_060047.3 | ||||
XM_005247546.2 | Intron | XP_005247603.1 | ||||
XM_005247547.1 | Intron | XP_005247604.1 | ||||
XM_011512930.1 | Intron | XP_011511232.1 | ||||
XM_011512931.1 | Intron | XP_011511233.1 | ||||
XM_017006646.1 | Intron | XP_016862135.1 | ||||
XM_017006647.1 | Intron | XP_016862136.1 | ||||
XM_017006648.1 | Intron | XP_016862137.1 | ||||
XM_017006649.1 | Intron | XP_016862138.1 |